Anti-NAK ATPASE Mouse Monoclonal Antibody

Supplier: Genetex
GTX30202
89362-888EA 735.54 USD
89362-888
Anti-NAK ATPASE Mouse Monoclonal Antibody
Antibodies
Na,K-ATPase is an integral membrane protein complex that hydrolyzes ATP to maintain the transmembrane gradients of Na+ and K+ found in most mammalian cells. The enzyme is comprised of an alpha and beta subunit. The alpha-polypeptide has been shown to be the catalytically active subunit, wherease the Beta-polypeptide appears to be necessary for the assembly and transport of the sodium pump to the plasma membrane. Na,K-ATPase alpha-1 subunit, in brain, is expressed in both neuronal and non-neuronal cells.

Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR