Anti-PRKCG Rabbit Polyclonal Antibody

Supplier: Genetex
GTX25778
89361-730EA 620.96 USD
89361-730
Anti-PRKCG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PKC gamma ( phospho T514 )

WB: Use at a concentration of 0.5 - 1.0 ?g/ml. Detects a band of approximately 80 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRKCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR