Anti-HIV1 P24 Mouse Monoclonal Antibody [clone: 473]

Supplier: Genetex
GTX19054
89360-548EA 622.3 USD
89360-548
Anti-HIV1 P24 Mouse Monoclonal Antibody [clone: 473]
Antibodies
HIV1 performs highly complex orchestrated tasks during the assembly, budding, maturation, and infection stages of the viral replication cycle. During viral assembly, the proteins form membrane associations and self-associations that ultimately result in budding of an immature virion from the infected cell. Gag precursors also function during viral assembly to selectively bind and package two plus strands of genomic RNA.

For ELISA: Use at an assay dependent dilution. Reactivity against HIV1 >10-5. For WB: Use at a concentration of 0.1-1 ?g/ml. This concentration will allow visualization of 100ng/lane of recombinant HIV1 p24. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: HIV1 P24
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR