Anti-RNA Polymerase beta Mouse Monoclonal Antibody

Supplier: Genetex
GTX12087
89359-166EA 612.56 USD
89359-166
Anti-RNA Polymerase beta Mouse Monoclonal Antibody
Antibodies
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.

Type: Primary
Antigen: RNA Polymerase beta
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR