Anti-RIP Rabbit Polyclonal Antibody

Supplier: Genetex
GTX111074
89356-554EA 594.42 USD
89356-554
Anti-RIP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RIP (receptor (TNFRSF)-interacting serine-threonine kinase 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR