Anti-TC21 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX110559
89356-368EA 594.42 USD
89356-368
Anti-TC21 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TC21 (related RAS viral (r-ras) oncogene homolog 2)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TC21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR