Anti-NRAS Rabbit Polyclonal Antibody

Supplier: Genetex
GTX108598
89355-612EA 594.42 USD
89355-612
Anti-NRAS Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to N-Ras (neuroblastoma RAS viral (v-ras) oncogene homolog)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR