Anti-TRF1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX106349
89354-638EA 594.42 USD
89354-638
Anti-TRF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TRF1 (telomeric repeat binding factor (NIMA-interacting) 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR