Anti-PPP1R16A Rabbit Polyclonal Antibody

Supplier: Genetex
GTX105410
89354-242EA 594.42 USD
89354-242
Anti-PPP1R16A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PPP1R16A (protein phosphatase 1, regulatory (inhibitor) subunit 16A)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PPP1R16A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR