Anti-SUGT1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX105358
89354-218EA 594.42 USD
89354-218
Anti-SUGT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SUGT1 (SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SUGT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR