Anti-CDC7 Mouse Monoclonal Antibody [clone: DCS-341]

Supplier: Genetex
GTX10535
89354-210EA 669.76 USD
89354-210
Anti-CDC7 Mouse Monoclonal Antibody [clone: DCS-341]
Antibodies
Mouse monoclonal [DCS - 341] to Cdc7 Kinase

ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a concentration of 0.4 µg/ml. Detects a band of approximately 55 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CDC7
Clonality: Monoclonal
Clone: DCS-341
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR