Anti-FN1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX72724
89353-052EA 664.02 USD
89353-052
Anti-FN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Fibronectin

Immunohistochemistry (IHC) 1:100-1:200 for 30 minute at room temperature using polymer system or streptavidin-biotin system. Formalin-fixed paraffin-embedded (FFPE) tissue sections and cell smears require antigen retriever, boiling tissue in 10 mM citrate buffer, pH 6.0 for 10-20 minute, followed by cooling at room temperature for 20 minutes.

Type: Primary
Antigen: FN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR