Anti-KRT15 Mouse Monoclonal Antibody [clone: LHK15]

Supplier: Genetex
GTX72325
89352-992EA 365.48 USD
89352-992
Anti-KRT15 Mouse Monoclonal Antibody [clone: LHK15]
Antibodies
Mouse monoclonal antibody [LHK15] to Cytokeratin 15

For IF: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KRT15
Clonality: Monoclonal
Clone: LHK15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR