Anti-CD3 Mouse Monoclonal Antibody [clone: F7.2.38]

Supplier: Genetex
GTX17143
89351-338EA 1067.08 USD
89351-338
Anti-CD3 Mouse Monoclonal Antibody [clone: F7.2.38]
Antibodies
This antibody labels CD3 epsilon and is a useful tool for the identification of T cells and related neoplasms

IHC (formalin/PFA fixed paraffin-embedded sections): Use at 1:10 - 1:25. We suggest an incubation period of 30-60 minutes at room temperature. Perform heat mediated antigen retrieval before commencing with IHC staining protocol. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR