Anti-CK5 Mouse Monoclonal Antibody [clone: D5/16 B4]

Supplier: Genetex
GTX17133
89351-336EA 626.93 USD
89351-336
Anti-CK5 Mouse Monoclonal Antibody [clone: D5/16 B4]
Antibodies
Cytokeratin 5 (58 kD) is a high molecular weight, basic type of cytokeratin expressed in basal, the intermediate and the superficial cell layers of stratified epithelia as well as transitional epithelia, complex epithelia and in mesothelial cells and mesothelioma. Cytokeratin 6 (56 kD) is also a high molecular weight, basic type cytokeratin expressed by proliferating squamous epithelium often paired with Cytokeratin 16.

IHC-P: 1/25 - 1/50. This dilution is suitable to use with the ABC method. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CK5
Clonality: Monoclonal
Clone: D5/16 B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR