Anti-EBV Mouse Monoclonal Antibody

Supplier: Genetex
GTX17085
89351-316EA 623.9 USD
89351-316
Anti-EBV Mouse Monoclonal Antibody
Antibodies
This product is a cocktail of monoclonal antibodies.

Type: Primary
Antigen: EBV
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR