Anti-MUC16 Mouse Monoclonal Antibody [clone: X75]

Supplier: Genetex
GTX10029
89350-588EA 575.15 USD
89350-588
Anti-MUC16 Mouse Monoclonal Antibody [clone: X75]
Antibodies
Mouse monoclonal antibody [X75] to CA125

For ELISA: Use at an assay dependent dilution. For IHC-P: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: X75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR