Anti-RSV Mouse Monoclonal Antibody [clone: 9C5]

Supplier: Genetex
GTX10018
89350-576EA 710.1 USD
89350-576
Anti-RSV Mouse Monoclonal Antibody [clone: 9C5]
Antibodies
Mouse monoclonal [9C5] to Respiratory Syncytial Virus

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 9C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Respiratory syncytial virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR