Anti-POLR2B Rabbit Polyclonal Antibody

Supplier: Genetex
GTX102535
89349-620EA 594.42 USD
89349-620
Anti-POLR2B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RNA polymerase IIB (DNA-directed RNA polymerase II subunit RPB2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: POLR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR