Anti-BRAF Rabbit Polyclonal Antibody

Supplier: Genetex
GTX100913
89348-752EA 594.42 USD
89348-752
Anti-BRAF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to B-Raf (v-raf murine sarcoma viral oncogene homolog B1)

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BRAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR