Anti-C-YES Rabbit Polyclonal Antibody

Supplier: Genetex
GTX100616
89348-568EA 594.42 USD
89348-568
Anti-C-YES Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to c-Yes (v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: C-YES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR