Anti-ATF2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX100224
89348-296EA 594.42 USD
89348-296
Anti-ATF2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ATF2 (activating transcription factor 2)

  • Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR