Fetal Bovine Serum, U.S. Origin, Genetex
Supplier: Genetex
GTX73252
89347-790EA 1331.69 USD
89347-790
Fetal Bovine Serum, U.S. Origin, Genetex
Sera (Serum)
FBS is ideal for use in ELISA, western blotting, immunohistochemistry, immunofluoroscence and other applications.
The serum has to be stored at –80 °C or below. Freeze-thaw cycles are to be avoided and standard sterile procedures are to be handled to maintain it's sterility.
Caution: For In vitro laboratory use only. Not for any clinical, therapeutic, or diagnostic use in humans or animals. Not for animal or human consumption.