Anti-CD11b (Integirn alpha M) Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M1/70.15]

Supplier: Genetex
GTX82969
89337-340EA 667.99 USD
89337-340
Anti-CD11b (Integirn alpha M) Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M1/70.15]
Antibodies
Rat monoclonal [M1/70.15] to mouse CD11b (FITC)

For Blocking: Use at an assay dependent dilution. For FACS: Use at an assay dependent dilution. For IHC-Fr: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD11b (Integirn alpha M)
Clonality: Monoclonal
Clone: M1/70.15
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR