Anti-DNMT3B (DNA methyltransferase 3B) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX125917
89336-016EA 778.77 USD
89336-016
Anti-DNMT3B (DNA methyltransferase 3B) Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DNMT3B (DNA methyltransferase 3B)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR