Anti-BEST3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX106048
89335-412EA 594.42 USD
89335-412
Anti-BEST3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HSFY1+2 (heat shock transcription factor, Y-linked 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BEST3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR