Anti-RBBP9 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX102333
89335-352EA 594.42 USD
89335-352
Anti-RBBP9 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RelB (v-rel reticuloendotheliosis viral oncogene homolog B)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, IP: 1:100-1:500, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RBBP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR