Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: 6581]

Supplier: Genetex
GTX36721
89331-800EA 665.25 USD
89331-800
Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: 6581]
Antibodies
Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: Monoclonal
Clone: 6581
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR