Anti-FNBP1L Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85425
89329-500EA 620.88 USD
89329-500
Anti-FNBP1L Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TOCA-1

Type: Primary
Antigen: FNBP1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR