Anti-DLGAP3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85421
89329-492EA 620.88 USD
89329-492
Anti-DLGAP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SAPAP3

Type: Primary
Antigen: DLGAP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR