Anti-CAPS2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85405
89329-460EA 620.88 USD
89329-460
Anti-CAPS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CAPS2

Type: Primary
Antigen: CAPS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR