Anti-JPH2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85367
89329-386EA 641.04 USD
89329-386
Anti-JPH2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to JPH2

Type: Primary
Antigen: JPH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR