Anti-APPL1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX114648
89321-780EA 594.42 USD
89321-780
Anti-APPL1 Rabbit Polyclonal Antibody
Antibodies
  • Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: APPL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR