Anti-MBNL1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX113264
89321-218EA 594.42 USD
89321-218
Anti-MBNL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to REL (v-REL reticuloendotheliosis viral oncogene homolog (avian))

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MBNL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR