Anti-OR8I2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX110506
89320-536EA 594.42 USD
89320-536
Anti-OR8I2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Apc11 (APC11 anaphase promoting complex subunit 11 homolog (yeast))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR8I2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR