Anti-LRP10 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX110405
89320-520EA 594.42 USD
89320-520
Anti-LRP10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to APBB3 (amyloid beta (A4) precursor protein-binding, family B, member 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LRP10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR