Anti-CEP97 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX110309
89320-502EA 655.77 USD
89320-502
Anti-CEP97 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Lamin B2 (lamin B2)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CEP97
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR