Anti-UGDH Rabbit Polyclonal Antibody

Supplier: Genetex
GTX109941
89320-434EA 594.42 USD
89320-434
Anti-UGDH Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PNPLA2 (patatin-like phospholipase domain containing 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: UGDH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR