Anti-GFAP Rabbit Polyclonal Antibody

Supplier: Genetex
GTX104790
89319-270EA 594.42 USD
89319-270
Anti-GFAP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Asporin (asporin)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ASPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR