Anti-GGPS1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX104786
89319-268EA 594.42 USD
89319-268
Anti-GGPS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SIPA1 (signal-induced proliferation-associated 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SIPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR