Anti-MDM4 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX101222
89318-380EA 594.42 USD
89318-380
Anti-MDM4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MDM4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR