Anti-FRA2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX15296
89317-928EA 616.98 USD
89317-928
Anti-FRA2 Rabbit Polyclonal Antibody
Antibodies
IHC-P: Use at a dilution of 1/50 for 30 minutes at RT. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10 minutes followed by cooling at RT for 20 minutes. WB: Use at an assay dependent dilution. Predicted molecular weight: 36 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FOSL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR