Anti-CR2 Mouse Monoclonal Antibody [clone: LT21]

Supplier: Genetex
GTX21090
89316-116EA 645.16 USD
89316-116
Anti-CR2 Mouse Monoclonal Antibody [clone: LT21]
Antibodies
FACS: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: LT21
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR