Anti-IgG Mouse Monoclonal Antibody [clone: IVA285-1]

Supplier: Genetex
GTX78382
89315-886EA 382.35 USD
89315-886
Anti-IgG Mouse Monoclonal Antibody [clone: IVA285-1]
Antibodies
Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone: IVA285-1
Conjugation:
Epitope: Light chain
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR