Anti-IgG2c Mouse Monoclonal Antibody

Supplier: Genetex
GTX35043
89306-986EA 135.53 USD
89306-986
Anti-IgG2c Mouse Monoclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG2c
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR