Anti-FKHRL1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX50856
89306-852EA 650.41 USD
89306-852
Anti-FKHRL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FOXO3A

Suggested starting dilutions are as follows. WB: 1:500~1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FOXO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR