Anti-MAPKK5 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX50845
89306-830EA 625.61 USD
89306-830
Anti-MAPKK5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MEK5(Phospho Ser311/ Thr315)

Suggested starting dilutions are as follows. WB: 1:500~1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAPKK5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR