Anti-c-Kit (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX50271
89304-398EA 625.61 USD
89304-398
Anti-c-Kit (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog) Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to c-Kit (Phospho Tyr721)

Suggested starting dilutions are as follows. WB: 1:500~1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: c-Kit (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR