Anti-HSV1 Mouse Monoclonal Antibody [clone: B323M]

Supplier: Genetex
GTX41420
89303-508EA 987.03 USD
89303-508
Anti-HSV1 Mouse Monoclonal Antibody [clone: B323M]
Antibodies
Mouse Monoclonal antibody [B323M] to HSV-1

Type: Primary
Antigen: HSV1
Clonality: Monoclonal
Clone: B323M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR