Anti-TNK1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX81327
89301-744EA 622.81 USD
89301-744
Anti-TNK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to TNK1 (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000IHC: Use at 1:50-100WB: Use at 1:250-500Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: TNK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR